Learn More
Invitrogen™ FM™ 1-43 Dye (N-(3-Triethylammoniumpropyl)-4-(4-(Dibutylamino) Styryl) Pyridinium Dibromide)
Membrane probe
£514.00 - £600.00
Specifications
Content And Storage | Store at room temperature and protect from light. |
---|---|
For Use With (Equipment) | Fluorescence Microscope |
Detection Method | Fluorescent |
Product Line | FM™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10769623
|
Invitrogen™
T3163 |
1mg |
£514.00
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11500786
|
Invitrogen™
T35356 |
10 x 100μg |
£600.00
1000µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FM 1-43 is an excellent reagent both for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling. This water-soluble dye, which is nontoxic to cells and virtually nonfluorescent in aqueous medium, is believed to insert into the outer leaflet of the cell membrane where it becomes intensely fluorescent. In a neuron that is actively releasing neurotransmitters, the dye becomes internalized within the recycled synaptic vesicles and the nerve terminals become brightly stained. The nonspecific staining of cell-surface membranes can simply be washed off prior to viewing.
FM 1-43 membrane probe is also available as 1mg packaged into 10 vials, each containing 100μg (T-35356). FM 1-43FX membrane probe (F-35355), an analog of FM 1-43 membrane probe that can be fixed in place using aldehyde-based fixatives, is also available.
Cell Analysis, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Endocytosis, Exocytosis & Phagocytosis, General Cell Tracing, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature and protect from light. | |
Fluorescent | |
Room Temperature | |
Dye |
Fluorescence Microscope | |
FM™ | |
Fluorescent Dye | |
Plasma Membrane |
For Research Use Only. Not for use in diagnostic procedures.