Learn More
Invitrogen™ FocalCheck™ Microspheres, 15 μm, fluorescent green/orange/dark-red ring stains
Fluorescent green/orange/dark-red ring stains
Brand: Invitrogen™ F7235
430.35 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
The sharp ring stains exhibited by the 15μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.
Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)
Specifications
Confocal Microscope Calibration | |
Orange, Dark Red, Green | |
Suspension Beads | |
0.5 mL | |
15 μm |
Store in refrigerator 2°C to 8°C and protect from light. | |
Microsphere | |
FocalCheck | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.