missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Formyl-Nle-Leu-Phe-Nle-Tyr-Lys, Fluorescein Derivative
The green-fluorescent conjugate can be used to investigate the fMLF receptor by fluorescence microscopy or flow cytometry
Brand: Invitrogen™ F1314
This item is not returnable.
View return policy
Description
Cell Analysis, Flow Cytometry
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescein Derivative | |
Room Temperature |
Fluorescence | |
Classic Dyes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Formyl-Nle-Leu-Phe-Nle-Tyr-Lys, Fluorescein Derivative > 1mg
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction