missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FR-31564 (Fosmidomycin, Sodium Salt)

Product Code. 11514806
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11514806

Brand: Invitrogen™ F23103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Blocks a pathway of isoprene synthesis

The phosphonate antibiotic FR-31564 (fosmidomycin) is an effective antimalarial agent that functions by blocking a mevalonate-independent pathway of isoprene synthesis. The antibiotic activity of FR-31564 is potentiated by glucose 1-phosphate. FR-31564 is also active against a variety of gram-negative bacteria.

Analysis of Bacteria, Analysis of Viruses, Fungi and Parasites, Antibiotic Binding Studies, Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Microbiological Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Shipping Condition Room Temperature
Molecular Formula C4H9NNaO5P
Physical Form Powder
Product Type Antibiotic

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.