Learn More
Invitrogen™ Gelatin From Pig Skin, Fluorescein Conjugate
Useful for study of gelatinases and collagenases
Brand: Invitrogen™ G13187
Code : 50
Additional Details : Weight : 0.00525kg
Description
Molecular Probes fluorescent conjugates of gelatin are designed for researchers studying not only collagen-binding proteins and collagen metabolism, but also for the study of gelatinases and collagenases (metalloproteins that digest collagen and gelatin).
- Label (Ex/Em): Fluorescein (approx. 499/519nm)
- Degree of labeling is listed on the product label (based on molecular weight of 100,000)
Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
5 mg | |
Pig | |
Classic Dyes | |
Lyophilized |
Fluorescein | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Gelatin |
For Research Use Only. Not for use in diagnostic procedures.