Learn More
Invitrogen™ Griess Reagent Kit, for nitrite quantitation
For nitrite quantitation
Brand: Invitrogen™ G7921
199.50 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.13000kg
Description
- Greiss reagent kit provides simple and well characterized colorimetric assay for nitrites, and nitrates that have been reduced to nitrites, with detection limit of about 100nM
Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Nitric Oxide Research, Nitro-Oxidative Stress
Order Info
Shipping Condition: Room Temperature
Safety and Handling
- Griess Reagent Kit, for nitrite quantitation
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Phosphoric acid, 4-aminobenzenesulfonicacid
For Research Use Only. Not for use in diagnostic procedures.