missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Guanosine 5'-O-(3-Thiotriphosphate), BODIPY™ FL Thioester, Sodium Salt (BODIPY™ FL GTP-γ-S, Thioester)

Product Code. 10663993
Click to view available options
Quantity:
50 μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10663993

Brand: Invitrogen™ G22183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fluorescence may be recovered on binding to G-proteins

BODIPY FL GTP-γ-S binds to GTP-binding proteins. To create this fluorescent nucleotide, the BODIPY FL fluorophore was linked through the γ-thiol of GTP-γ-S. The fluorescence of BODIPY FL GTP-γ-S is quenched approx. 90% relative to that of the free dye, but may be recovered on binding to G-proteins. In addition to its potential use for binding studies, the BODIPY FL GTP-γ-S thioether is an important new substrate for Fhit, a member of the histidine triad superfamily of nucleotide-binding proteins.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Guanosine 5'-O
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Labeling Method Chemical Labeling, Direct Labeling
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye BODIPY™ FL
Quantity 50 μL
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.