Learn More
Invitrogen™ Guanosine 5'-Triphosphate, BODIPY™ FL 2'-(or-3')-O-(N-(2-Aminoethyl) Urethane), Trisodium Salt (BODIPY™ FL GTP)
Based on interactions between the fluorophore and the purine base
Brand: Invitrogen™ G12411
293.55 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Descrizione
BODIPY FL adenosine 5'-triphosphate (A-12410) and BODIPY FL guanosine 5'-triphosphate are composed of a BODIPY FL fluorophore attached to the 2' or 3'' position of the ribose ring via a linker. Interactions between the fluorophore and the purine base are evident from the spectroscopic properties of these nucleotide analogs. The fluorescence quantum yield of BODIPY FL GTP is only about one-tenth that of BODIPY FL ATP; however, this yield may be enhanced upon binding of BODIPY FL GTP to GTP-binding proteins.
Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Order Info
Shipping Condition: Wet ice
Specifica
Guanosine 5'-Triphosphate | |
Wet Ice | |
BODIPY™ FL | |
BODIPY™ |
Store in freezer -5°C to -30°C and protect from light. | |
5 mM | |
100 μL |
For Research Use Only. Not for use in diagnostic procedures.