Learn More
Invitrogen™ Heparin, Fluorescein Conjugate
Shows heparin binding in cells and tissue
Brand: Invitrogen™ H7482
231.80 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The green-fluorescent fluorescein heparin conjugate should be a useful tool for studying binding of this mucopolysaccharide in cells and tissue.
Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors
Order Info
Shipping Condition: Room temperature
Specifications
1 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Other Proteins |
Fluorescein | |
Classic Dyes | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.