missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPTS; pyranine (8-Hydroxypyrene-1,3,6-Trisulfonic Acid, Trisodium Salt)
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
- pKa of approximately 7.3 in aqueous buffers
Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Ionic Homeostasis and Signaling, pH Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Color | Green |
| Content And Storage | Store at room temperature and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | pH Indicator |
| Quantity | 1 g |
| Shipping Condition | Room Temperature |
| Sub Cellular Localization | Cytoplasm & Cytosol |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction