Learn More
Invitrogen™ HybriSlip™ Hybridization Cover, 22 mm x 22 mm, RNase free
Designed specifically for in situ hybridization
Brand: Invitrogen™ H18200
Code : 50
Additional Details : Weight : 0.00450kg
Description
- Does not require lengthy preparation or blocking procedures to prevent probe trapping or binding
- Heat resistant and do not curl, even at high temperatures
- Well-suited for denaturation steps or in situ PCR incubations
Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Fluorescence In Situ Hybridization, In Situ Hybridization (ISH)
Order Info
Shipping Condition: Room temperature
Specifications
Hybridization Cover | |
22 x 22 mm | |
1 set |
HybriSlip | |
Store at room temperature. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.