missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Escherichia coli (K-12 strain) BioParticles™, Texas Red™ conjugate
The fluorescence of the dye conjugates is uniformly intense over the pH range from 3 to 10.
Brand: Invitrogen™ E2863
165.00 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Conjugates covalently labeled with variety of different fluorophores (special care has been taken to remove free dye after conjugation)
- Provided as lyophilized powders
- Approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
- Conjugates can be reconstituted in the buffer of choice for use in phagocytosis assays
- Fluorescence of conjugates that are bound to surface of cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
- Addition to cellular applications, fluorescent conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
- Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
Mammalian | |
Fluorescence | |
E.coli Conjugate | |
Solid | |
10 mg | |
E. coli |
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence Microscope | |
Texas Red™ | |
BioParticles™, Texas Red™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.