Learn More
Invitrogen™ Image-IT™ DEAD Green™ Viability Stain
Stain becomes permeant when plasma membrane is damaged
Brand: Invitrogen™ I10291
Code : 50
Additional Details : Weight : 0.10000kg
Includes: The stain includes sufficient material to perform 10 plates in a 96-well plate format and 50μL well volume.
Description
Image-iT DEAD Green viability stain is a fast, sensitive and easy-to-use viability indicator that is also amenable to fixation and permeabilization, thus allowing for multiplexing with other biomarkers of cytotoxicity. Image-iT DEAD Green is an impermeant dye to healthy cells that becomes permeant when the plasma membrane of cells is compromised.
Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function
Order Info
Shipping Condition: Room temperature
Specifications
Green | |
The stain includes sufficient material to perform 10 plates in a 96-well plate format and 50 μL well volume. |
|
Viability Assay | |
Viability Stain | |
Liquid | |
Wet Ice |
1 mM | |
Fluorescent | |
Fluorescence Microscope | |
Visible | |
Image-iT™, Molecular Probes™ | |
Nucleus |
For Research Use Only. Not for use in diagnostic procedures.