missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ FX Signal Enhancer

Apply directly to slides or coverslips containing fixed and permeabilized cell or tissue samples prior to staining with fluorescent probes

Brand:  Invitrogen™ I36933

Product Code. 11584896

  • £105.00 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

When Image-iT FX signal enhancer is used, nonspecific fluorescence (background) commonly seen with the application of fluorescent conjugates of streptavidin, goat anti-mouse, or goat anti-rabbit IgG is largely eliminated.

  • Easy protocol—apply enhancer from ready-made solution in dropper bottle
  • Effective with many fluorescent labels
  • Cost effective—supplies enough for 50 standard coverslips
  • Compatible with immunostaining protocol's other blocking steps
  • Reduce Nonspecific Binding of Dyes to Tissues

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Epitope Retrieval, Blocking and Quenching

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Signal Enhancer
Signal Enhancer
Image-iT™
10 mL
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2896700 Certificate TypeCertificate of Analysis Date03/04/2024 Catalog NumberI36933
Lot Number2795285 Certificate TypeCertificate of Analysis Date19/11/2023 Catalog NumberI36933
Lot Number2622403 Certificate TypeCertificate of Analysis Date28/04/2023 Catalog NumberI36933
Lot Number2506074 Certificate TypeCertificate of Analysis Date03/07/2022 Catalog NumberI36933
Lot Number2421200 Certificate TypeCertificate of Analysis Date13/12/2021 Catalog NumberI36933
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Image-iT™ FX Signal Enhancer > 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.