missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ Lipid Peroxidation Kit, for live cell analysis

Enables detection of lipid peroxidation in live cells through oxidation of BODIPY™ 581/591 C11 reagent

Brand:  Invitrogen™ C10445

313.12 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.



Product Code. 12877552

  • £412.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Five single-use vials with enough total reagent for five 96-well plates or 100 coverslips, and cumene hydroperoxide as a positive control
This item is not returnable. View return policy

Description

Description

This kit localizes to membranes throughout live cells and upon oxidation by lipid hydroperoxides, displays a shift in peak fluorescence emission from approx. 590nm to approx. 510nm. Fluorescence from live cells shifts from red to green, providing a ratiometric indication of lipid peroxidation compatible with traditional and high content microscopy, as well as flow cytometry.

  • Includes cumene hydroperoxide as positive control compound to induce lipid peroxidation in cells
  • Reagent is provided as stable, ready-to-use DMSO solution with a simple protocol compatible with standard workflows in fluorescence microscopy
  • Form: Frozen

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress, Reactive Oxygen Species

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
Frozen
1 kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Image-iT Lipid Peroxidation Kit, for live cell analysis

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Organic peroxide Type E & F
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H242-Heating may cause a fire.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Cumene hydroperoxide, Dimethyl sulfoxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.