Learn More
Invitrogen™ Image-IT™ LIVE Green Reactive Oxygen Species Detection Kit, for microscopy
For microscopy
Brand:  Invitrogen™ I36007
364.04 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10738994
  • £479.00 / 1 set
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
  • Image-iT LIVE Green Reactive Oxygen Species (ROS) Detection Kit provides key reagents necessary for detection of ROS in live cells
  • Assay is based on 5-(and-6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate (carboxy-H2DCFDA), reliable fluorogenic marker for ROS in live cells
  • In addition to carboxy-H2DCFDA, kit provides common inducer of ROS production tert-butyl hydroperoxide (TBHP), as positive control and blue-fluorescent, cell-permeant nucleic acid stain Hoechst 33342
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 kit