missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ InSpeck™ Green (505/515) Microscope Image Intensity Calibration Kit, 6 μm

Microspheres have diameters close to those of many stained cells, providing fluorescence microscopists with intensity references for generating calibration curves and evaluating instrument performance

Brand:  Invitrogen™ I14785

Product Code. 10533483

  • £266.00 / 1 set
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Aqueous suspension of microspheres can be directly applied to sample for calibrating fluorescence intensities, or they can be mounted separately in adjacent well or on another slide

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Microscope Image Intensity Calibration Kit
Microscope Image Intensity Calibration
Suspension Beads
Store in refrigerator 2°C to 8°C and protect from light.
Room Temperature
InSpeck
Green
6 μm
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ InSpeck™ Green (505/515) Microscope Image Intensity Calibration Kit, 6 μm > Green; 6um

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.