Learn More
Invitrogen™ Latrunculin A
For applications involving cell structure
Brand: Invitrogen™ L12370
Code : 50
Additional Details : Weight : 0.00525kg
Description
Latrunculin A disrupts microfilament organization in cultured cells by binding to monomeric G-actin in a 1:1 complex at submicromolar concentrations. Its physiological effects include inhibition of fertilization, early embryological development and phagocytic internalization of immune complexes.
Cell Analysis, Cell Structure, Cytoskeleton
Order Info
Shipping Condition: Room Temperature
Specifications
Room Temperature | |
C22H31NO5S | |
Inhibitor |
Actin, Cytoskeleton | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.