Learn More
Invitrogen™ Lectin PHA-L From Phaseolus vulgaris (red kidney bean), Alexa Fluor™ 488 Conjugate
Suitable for use in cell tracing and tracking, cellular imaging and flow cytometry
Brand: Invitrogen™ L11270
Code : 50
Additional Details : Weight : 0.00525kg
Description
Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues.[ALTREF PN29369] This lectin has been shown to agglutinate leukocytes and induce mitogenic activity in T-lymphocytes[ALTREF PN29369,PN29368] and has also been used as an anterograde tracer for the study of neuronal pathways.[ALTREF PN20284]
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence Staining & Detection, Microbial Tracking
Order Info
Shipping Condition: Room Temperature
Specifications
1 mg | |
Alexa Fluor 488 | |
Alexa Fluor Dyes | |
Lyophilized | |
Alexa Fluor™ |
Red kidney bean | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Lectins |
For Research Use Only. Not for use in diagnostic procedures.