Learn More
Invitrogen™ HCS LipidTOX™ Red Phospholipidosis Detection Reagent (1000X), for cellular imaging
Characterizes side effects of certain compounds on lipid metabolism in mammalian cell lines
Brand: Invitrogen™ H34351
Code : 50
Additional Details : Weight : 0.50000kg
Description
The HCS LipidTOX Red phospholipidosis detection reagent was developed to characterize the potentially toxic side effects of compounds on lipid metabolism in mammalian cell lines. This reagent is easily detected by fluorescence microscopy or an HCS reader equipped with filters that can detect excitation/emission maxima approx. 595/615nm. LipidTOX phospholipidosis detection reagents do not affect the normal growth of cells, and the staining observed after incubation in live cells is maintained after formaldehyde fixation. This probe is compatible with the HCS LipidTOX neutral lipid stains.
Cell Analysis, Cellular Imaging, High-Content Screening (HCS)
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
High Content Analysis Instrument | |
Other Label(s) or Dye(s) | |
LipidTOX™ |
Fluorescent | |
Phospholipidosis Detection Reagent | |
96-well plate | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.