Learn More
Invitrogen™ Lissamine™ Rhodamine B 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt (rhodamine DHPE)
With excitation/emission maxima ∽560/580nm
Brand: Invitrogen™ L1392
161.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
The phospholipid, rhodamine DHPE is labeled on the head group with the red-fluorescent rhodamine B fluorophore.
Cell Analysis, Cell Structure, Membranes (General) & Lipids
Order Info
Shipping Condition: Room Temperature
Specifications
5 mg | |
Solid | |
Invitrogen |
For Research Use Only. Not for use in diagnostic procedures.