Learn More
Invitrogen™ Lucifer Yellow CH, Lithium Salt, 25 mg
Lucifer yellow CH, lithium salt is a water-soluble dye with excitation/emission peaks of 428/536 nm.
Brand: Invitrogen™ L453
152.57 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Description
Lucifer yellow CH, lithium salt is a water-soluble dye with excitation/emission peaks of 428/536 nm. It is a favorite tool for studying neuronal morphology, because it contains a carbohydrazide (CH) group that allows it to be covalently linked to surrounding biomolecules during aldehyde fixation. The lithium salt form of the lucifer yellow CH is commonly used for microinjection because it has higher solubility than the potassium and ammonium salt forms of lucifer yellow CH.
To make a concentrated stock solution, dissolve in H2O up to 8% (w/v). Store frozen and protected from light.
Order Info
Shipping Condition: Room temperature
Specifications
Yellow | |
Lithium Salt | |
Room Temperature | |
Cytoplasm & Cytosol | |
457.24 |
Fluorescence | |
Store at room temperature and protect from light. | |
Fluorescence Microscope | |
428/536 nm |
For Research Use Only. Not for use in diagnostic procedures.