Learn More
Invitrogen™ MANT-ATP (2'-(or-3')-O-(N-Methylanthraniloyl) Adenosine 5'-Triphosphate, Trisodium Salt)
Produces nucleotide analogs causing little perturbation of nucleotide-protein interactions
Brand: Invitrogen™ M12417
383.80 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Description
The nucleotide analog MANT ATP is modified on the ribose moiety. The compact mature of the MANT fluorophore and its attachment position results in nucleotide analogs that induce minimal perturbation of nucleotide-protein interactions. Because MANT fluorescence is sensitive to the environment of the fluorophore, nucleotide-protein interactions may be directly detectable. MANT nucleotides are valuable probes of the structure and enzymatic activity of nucleotide-binding proteins.
Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Order Info
Shipping Condition: Wet ice
Specifications
MANT-ATP | |
Wet Ice | |
MANT (N-Methylanthraniloyl) |
Store in freezer -5°C to -30°C and protect from light. | |
5 mM | |
400 μL |
For Research Use Only. Not for use in diagnostic procedures.