Learn More
Invitrogen™ Marina Blue™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Marina Blue™ DHPE)
A phospholipid
Brand: Invitrogen™ M12652
147.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
Marina Blue DHPE is labeled on the head group with the blue-fluorescent Marina Blue fluorophore with excitation/emission maxima ∽365/460nm.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Phospholipids | |
1 mg | |
Solid | |
MARINA BLUE |
For Research Use Only. Not for use in diagnostic procedures.