missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Methotrexate, Fluorescein, triammonium Salt (Fluorescein Methotrexate)

Product Code. 10655013
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10655013

Brand: Invitrogen™ M1198MP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

To probe DHFR expression

The green-fluorescent fluorescein methotrexate (absorption/emission maxima approx. 496/516nm) can be used to probe dihydrofolate reductase (DHFR) expression by fluorescence microscopy or flow cytometry.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Label Type Classic Dyes
Molecular Formula C46H54N14O9S
Physical Form Solid
Product Type Fluorescein Methotrexate
compliance-icons compliance-icons
Product Identifier
  • Methotrexate, Fluorescein, triammonium Salt (Fluorescein Methotrexate)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methotrexate, fluorescein, triammonium salt

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.