Learn More
Invitrogen™ MycoFluor™ Mycoplasma Detection Kit
Identify mycoplasma infection in cell cultures visually
Brand: Invitrogen™ M7006
254.60 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
The MycoFluor™ Mycoplasma Detection Kit provides an ultrasensitive, rapid, and simple fluorescence microscopic assay for the visual identification of mycoplasma infection in laboratory cell cultures. In order to detect mycoplasma, the fluorescent MycoFluor reagent is added directly to the culture medium, with or without cells, and the stained sample is then examined under a fluorescence microscope.
Bioproduction, Cell Analysis, Cell Culture, Cell Tracing and Tracking, Clinical, Contaminant and Impurity QC Testing, Infectious Disease Diagnostics, Mammalian Cell Culture, Microbial Tracking, Molecular Diagnostics, Mycoplasma Detection
Order Info
Shipping Condition: Room temperature
Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Liquid | |
Fluorescence Microscope | |
Room Temperature |
Fluorescence | |
Mycoplasma Detection | |
100 tests |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.