Learn More
Invitrogen™ NanoOrange™ Protein Quantitation Kit, 200-2,000 assays
Sensitive and easy assay for protein quantitation, with detection as low as 10 ng/mL of protein in solution
Brand: Invitrogen™ N6666
411.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Fluorescent dye is suitable for use with spectrofluorometers and microplate readers
- For detection of lipoproteins or proteins in a complex lipid environment,use CBQCA Protein Quantitation Kit (C-6667)
Protein Quantitation, Proteins, Expression, Isolation and Analysis

Specifications
Fluorescent Protein Assay | |
NanoOrange™ | |
200 to 2000 Assays | |
Microplate Reader |
Fluorescence | |
Protein Quantitation Assay | |
Protein quantitation | |
1 Kit |
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.