Learn More
Invitrogen™ NBD C6-Ceramide (6-((N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)hexanoyl)Sphingosine)
Selective stain for Golgi apparatus in live and fixed cells
Brand:  Invitrogen™ N1154
277.40 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10226912
  • £365.00 / 1mg
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
NBD C6-ceramide can be used to study sphingolipid transport and metabolism mechanisms. The environment sensitive NBD dye is weakly fluorescent in water but increases in aprotic solvents and other nonpolar environments with excitation/emission maxima ∽466/536nm.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sphingolipids
1 mg
Solid
Invitrogen