missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NBT/BCIP Reagent Kit
For chromogenic detection of phosphatase.
Brand: Invitrogen™ N6547
289.56 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: 1g each of NBT and BCIP
Description
- NBT/BCIP Reagent Kit includes 1g samples of NBT (Nitro blue Tetrazolium Chloride) and BCIP (5-bromo-4-chloro-3-indolyl-1-phosphate)
Cell Analysis, Cellular Imaging, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunohistochemistry (IHC), Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
1 kit | |
Chemical Substrate |
Colorimetric | |
Phosphatase Substrate | |
Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction