Learn More
Invitrogen™ NeuroTrace™ BDA-10,000 Neuronal Tracer Kit
Contains neuronal tracer, biotin dextran amine (BDA), and detection reagents, avidin-horseradish peroxidase (avidin-HRP) and diaminobenzedine (DAB) for subsequent visualization by light or electron microscopy
Brand: Invitrogen™ N7167
Alert:
DAB is mutagenic.
Includes: The NeuroTrace BDA-10,000 Neuronal Tracer Kit contains convenient amounts of eachof the components required for neuroanatomical tracing withBDA, including; 10,000 MW lysine-fixable biotin dextran amine; avidin-horseradish peroxidase; 3,3'-diaminobenzidine
Description
The neuronal tracer biotin dextran amine (BDA) is transported over long distances and fills fine neuronal processes bidirectionally, including boutons in the anterograde direction and dendritic structures in the retrograde direction.1-3 Days to weeks after BDA is injected into the desired region of the brain, the brain tissue is fixed, sectioned and incubated with avidin-horseradish peroxidase (avidin-HRP). Following a diaminobenzidine (DAB) reaction, the electron-dense DAB reaction product can be viewed by either light or electron microscopy.
Cell Analysis, Cell Tracing and Tracking, Neuronal Tracing
Order Info
Shipping Condition: Wet Ice
Specifications
Colorimetric | |
1 kit | |
Wet Ice | |
Cytoplasm & Cytosol |
Neuronal Tracer Kit | |
Store in freezer (-5 to -30°C) and protect from light. | |
Electron Microscope, Light Microscope | |
NEUROTRACE™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.