missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucBlue™ Fixed Cell ReadyProbes™ Reagent (DAPI)

High-purity form of DAPI stain in room temperature-stable solution that is provided in dropper bottle

Brand:  Invitrogen™ R37606

113.24 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 12333553

  • £149.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

DAPI is a commonly used nuclear counterstain for fixed cells that emits blue fluorescence when bound to DNA.

  • In fluorescence microscopy, DAPI is excited with UV light and detected through blue/cyan filter
  • DAPI is typically supplied as solid that requires weighing out and diluting several thousand-fold before use, and recommended storage is generally in freezer or refrigerator
  • This version is ready to use and can be stored next to microscope or workstation
  • Spectral properties of DAPI (4′,6-diamidino-2-phenylindole), its selectivity for dsDNA, and its increase in fluorescence when bound to DNA make it excellent fluorescent nuclear counterstain ideal for use with green (Alexa Fluor 488, FITC, GFP) and red (Alexa Fluor 594, rhodamine, Texas Red, mCherry, mKate-2) fluorophores in multicolor experiments
  • Because of its high affinity to DNA, it is also frequently used for counting cells, measuring apoptosis, sorting cells based on DNA content, and as nuclear segmentation tool in high content imaging analysis
  • In most cases 2 drops/mL and an incubation of 15 to 30 minutes will produce bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications
  • Excited by UV light at 360nm when bound to DNA, with an emission maximum at 460nm
  • It is detected through blue/cyan filter
  • As preferred blue nuclear stain in fixed cell imaging experiments, well-suited for use with antibody-based applications

Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry of Cellular Processes, Fluorescence In Situ Hybridization, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, In Situ Hybridization (ISH), Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
360⁄460
Fluorescence Microscope, Flow Cytometer
Fluorescence
Liquid
Room Temperature
Nucleic Acid Stain
6 × 2.5 mL dropper bottles

Store at ≤ 25°C

DAPI
6 vial(s) kit
Visible
Molecular Probes™
Fluorescent Dye
Nucleus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.