Learn More
Invitrogen™ O-Rings for Attofluor™ Cell Chamber
Replacement part for durable and practical coverslip holder for viewing live-cell specimens on upright or inverted microscopes
Brand: Invitrogen™ O14804
Code : 50
Additional Details : Weight : 0.00450kg
Description
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)
Order Info
Shipping Condition: Room temperature
Specifications
O-ring | |
Store at room temperature. | |
Room Temperature |
Attofluor | |
10 Pack |
For Research Use Only. Not for use in diagnostic procedures.