Learn More
Invitrogen™ Oregon Green™ 488 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Oregon Green™ 488 DHPE)
With excitation/emission maxima ∽501/526 nm
Brand: Invitrogen™ O12650
158.00 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
The phospholipid, Oregon Green 488 DHPE is labeled on the head group with the green-fluorescent Oregon Green fluorophore. This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∽4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
Cell Analysis, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, pH Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Phospholipids | |
1 mg | |
Solid | |
Oregon Green |
For Research Use Only. Not for use in diagnostic procedures.