Learn More
Invitrogen™ Ovalbumin, Alexa Fluor™ 647 Conjugate
Labeled with bright, photostable and pH-insensitive Alexa Fluor 647 dye
Brand: Invitrogen™ O34784
Code : 50
Additional Details : Weight : 0.00230kg
Description
The far-red fluorescent Alexa Fluor 647 ovalbumin conjugate is a protein with a relatively low molecular weight (∽45,000 daltons).
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging
Order Info
Shipping Condition: Room Temperature
Specifications
2 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Albumin Proteins |
Alexa Fluor 647 | |
Alexa Fluor Dyes | |
Lyophilized | |
Alexa Fluor™ |
For Research Use Only. Not for use in diagnostic procedures.