missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PED-A1 (N-((6-(2,4-DNP)Amino)Hexanoyl)-1-(BODIPY™ FL C5)-2-Hexyl-Sn-Glycero-3-Phosphoethanolamine) (Phospholipase A1 Selective Substrate)

Product Code. 10083183
Click to view available options
Quantity:
100 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10083183

Brand: Invitrogen™ A10070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

PED-A1-labeled phospholipid

Molecular Probes phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells.

  • Label (Ex/Em): PED-A1
  • Specificity: PLA1
  • Cleavage product: BODIPY™ FL C5
  • Lyophilized product can be dissolved in DMSO for use
  • Fluorescence is measured either in microplate assays or using flow cytometry
  • Cell-Permeant
Note: Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Solid
Product Line BODIPY
Quantity 100 μg
Substrate Phospholipase A1 selective substrate
Substrate Properties Lipid-Based Substrate
Target Enzyme Phospholipase
For Use With (Equipment) Flow Cytometer, Microplate Reader
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.