Learn More
Invitrogen™ Pepstatin A, BODIPY™ FL Conjugate
For the study of cathepsin D trafficking in cells by fluorescence microscopy
Brand: Invitrogen™ P12271
162.67 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
BODIPY™ FL pepstatin A, a green-fluorescent probe, which inhibits cathepsin D in vitro (IC50 approx. 50nM), targets cathepsin D within lysosomes of live cells. Initial studies with this new probe indicate that it is transported to the lysosomes of live cells via an endocytic pathway and that it can be competitively displaced by unlabeled pepstatin A.
Apoptosis, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Enzyme and Protein Activity Assays, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Room Temperature | |
BODIPY | |
Solid | |
Inhibitor |
For Research Use Only. Not for use in diagnostic procedures.