Learn More
Invitrogen™ PIP Strips™ Membranes
Identify proteins possessing phosphoinositide domains
Brand: Invitrogen™ P23751
542.64 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
PIP Strips membranes are designed for the identification of proteins possessing phosphoinositide domains and for analysis of their lipid-binding specificities. PIP Strips membranes facilitate the analysis of phosphoinositide protein interactions by protein-lipid overlay assays. Proteins may be detected using standard western blot procedures in conjunction with high-performance alkaline phosphatase- and horseradish peroxidase (HRP)-mediated signal generation systems.
Cell Analysis, Cell Structure, Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Store in refrigerator 2°C to 8°C and protect from light. | |
Membrane | |
10 strips |
Phospholipid Arrays | |
PIP Strips™ | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.