Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Blot Stain Kit
Can be used to analyze the native phosphorylation state of proteins obtained from tissue specimens or from body fluids
Brand:  Invitrogen™ P33356
Product Code. 10378622
  • £352.00 / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
The Pro-Q Diamond Phosphoprotein Blot Stain Kit provides a rapid and simple method for directly detecting phosphoproteins on polyvinylidene difluoride (PVDF) or nitrocellulose membranes. The fluorescent stain binds selectively to the phosphate moiety and binding is independent of the amino acid sequence context.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
In-Blot Detection
Pro-Q Diamond
PRO-Q™
Proteins (Phosphoproteins)
Fluorescence
Phosphoprotein Blot Stain Kit
1 kit