Learn More
Invitrogen™ Molecular Probes™ ProLong™ Antifade Kit
Allows longer-term storage of the sample
Brand:  Invitrogen™ P7481
Product Code. 10720764
Description
Description
The ProLong Antifade Kit provides a powdered antifade reagent and a liquid mounting medium that can be mixed together and applied directly to fluorescently labeled cell or tissue samples on microscope slides to protect the dyes from fading (photobleaching) during fluorescence microscopy experiments.
  • Protects dyes from fading during imaging
  • Mix powdered antifade with mounting medium and use the same day
  • Mounted samples are stable for months
  • Offers excellent photobleaching protection for Texas Red™ and most Alexa Fluor™ dyes
  • Liquid mounting medium component can be used without antifade reagent for acceptor photobleaching FRET experiments
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer (-5 to -30°C) and protect from light.
1 kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Molecular Probes™ ProLong™ Antifade Kit > 1 kit