Learn More
Invitrogen™ β-Py-C10-HPC (1-Hexadecanoyl-2-(1-Pyrenedecanoyl)-sn-Glycero-3-Phosphocholine)
Phospholipid analog of phosphocholineβ-py-C10-HPC
Brand: Invitrogen™ H361
Code : 50
Additional Details : Weight : 0.00525kg
Description
Labeled with pyrene on the sn-2 acyl chain. Pyrene forms excimers at high concentrations, excimer emission ∽470nm. Pyrene is environment sensitive, essentially nonfluorescent in water.
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Phospholipids | |
1 mg | |
Invitrogen |
For Research Use Only. Not for use in diagnostic procedures.