missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ Protein Assay Kit

For easy and accurate protein quantitation

Brand:  Invitrogen™ Q33210

408.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10758463

  • £544.00 / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Kit provides concentrated assay reagent, dilution buffer, and pre-diluted BSA standards
  • Simply dilute reagent, load into wells of microplate, add 1-20μL of sample, mix, and measure fluorescence
  • Assay is highly selective for protein and exhibits little protein-to-protein variation
  • Assay is performed at room temperature, and signal is stable for 3hours
  • Common contaminants, such as salts, solvents, or DNA—but not detergents—are well tolerated in the assay
  • Quant-iT DNA Assay Kits (Q33120, Q33130) and a Quant-iT RNA Assay Kit (Q33140) are also available

Protein Quantitation, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C) and protect from light.
Fluorescence
Protein Quantitation Assay
3919448
1 Kit
Room Temperature
Fluorescent Protein Assay
Quant-iT™
Microplate Reader
Concentrated assay reagent, dilution buffer, and pre-diluted BSA standards
1 to 20 uL
Store in refrigerator (2° to 8°C). Protect from light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.