Learn More
Invitrogen™ Red/Green BODIPY™ PC-A2 (1-O-(6-BODIPY™ 558/568-Aminohexyl)-2-BODIPY™ FL C5-Sn-Glycero-3-Phosphocholine) (Ratiometric Phospholipase A2 Substrate)
Measure results in microplate assays or using flow cytometry
Brand: Invitrogen™ A10072
202.16 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Molecular Probes phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells. Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.
- Label (Ex/Em): Red/Green BODIPY™ PC-A2 (approx. 488/530,590nm) (can be used in dual emission fluorescence ratio detection)
- Specificity: PLA2
- Cleavage product: BODIPY FL C5
- Lyophilized product can be dissolved in DMSO for use
- Fluorescence is measured either in microplate assays or using flow cytometry
Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature

Specifications
Cell-Permeant | |
Fluorescence | |
BODIPY™ | |
Phospholipase A2 substrate (ratiometric) | |
Phospholipase | |
Room Temperature |
Store in freezer (-5 to -30°C). | |
Solid | |
100 μg | |
Lipid-Based Substrate | |
Flow Cytometer, Microplate Reader |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.