Learn More
Invitrogen™ Rhodamine 110, bis-(p-Tosyl-L-Glycyl-L-Prolyl-L-Arginine Amide)
This Molecular Probes™ bisamide derivative of rhodamine 110 (R110) is a sensitive and selective substrate for assaying proteases in solution or inside living cells.
Brand: Invitrogen™ R22124
307.34 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
322.70 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
This Molecular Probes™ bisamide derivative of rhodamine 110 (R110) is a sensitive and selective substrate for assaying proteases in solution or inside living cells. These fluorogenic substrates contain an amino acid or peptide covalently linked to each of R110's amino groups. Upon enzymatic cleavage, the nonfluorescent bisamide substrate is converted first to the fluorescent monoamide and then to R110, with a further increase in fluorescence. The substrate can be used to continuously measure enzyme activity in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader, or can be used for intracellular protease assays with analysis by flow cytometry or fluorescence microscopy.<
• Has spectral properties similar to those of fluorescein, making it compatible with standard fluorescein filter sets
• Signal to background ratios are good because of large extinction coefficient of R110
• Unlike fluorescein, the fluorescence intensity of the rhodamine 110 reaction product is constant from pH 3–9
• Substrate name: p-tosyl-Gly-Pro-Arg)2-R110
• Useful to measure: Thrombin
• Ex/Em of R110 product: (498/521nm)
• Cell-Permeant
Order Info
Shipping Condition: Room temperature

Specifications
Cell-Permeant | |
Fluorescence | |
2 mg | |
Peptide-Based Substrate | |
Fluorometer, Microplate Reader |
Store in freezer (-5 to -30°C). | |
Solid | |
Caspase Substrate | |
Thrombin | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.