Learn More
Invitrogen™ Secure-Seal™ Hybridization Chamber Gasket, 8 chambers, 9 mm diameter, 0.8 mm deep
Coverslip-seal combination that attaches to microscope slides to form microwells optimized for carrying out in situ hybridization procedures
Brand: Invitrogen™ S24732
Code : 50
Additional Details : Weight : 0.00450kg
Description
- Special adhesive that bonds to glass slides in seconds, creating water-tight seal that is temperature resistant but can also be removed cleanly and easily after hybridization
- Solutions are easily added or removed through dual-access ports without disrupting specimen
- Hydrophobic coverslips are nuclease-free and will not trap or bind probes, allowing uniform distribution of reagent over specimen
Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Fluorescence In Situ Hybridization, Gene Expression Analysis and Genotyping, In Situ Hybridization (ISH), Microarray Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Hybridization Chamber Gasket | |
0.8 mm | |
Store at room temperature. |
Secure-Seal | |
9 mm | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.