missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPQ (6-Methoxy-N-(3-Sulfopropyl)Quinolinium, Inner Salt)

Chloride indicator

Brand:  Invitrogen™ M440

188.10 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 11599096

  • £204.00 / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Absorption/emission maxima approximately 344/443nm
  • Detects Cl- via diffusion-limited collisional quenching

Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature and protect from light.
Confocal Microscope, Flow Cytometer
Fluorescent Dye-Based
Room Temperature
Fluorescence
Chloride Indicator
100 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.