missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTOX™ Green Nucleic Acid Stain - 5 mM Solution in DMSO

Useful indicator of dead cells within a population

Brand:  Invitrogen™ S7020

243.20 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10768273

  • £320.00 / 250µL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

SYTOX Green nucleic acid stain is an excellent green-fluorescent nuclear and chromosome counterstain that is impermeant to live cells.

Simple Viability Determination
SYTOX Green allows quick determination of cell viability when using flow cytometers, fluorescence microscopes, fluorometers, or fluorescence microplate readers. It will not cross intact membranes, but will easily penetrate compromised membranes characteristic of dead cells. Because it exhibits >500-fold fluorescence enhancement upon binding nucleic acids, briefly incubating dead cells with SYTOX Green will result in bright green fluorescence with an emission peak of 523nm when excited by a 488nm argon-ion laser or any other 450–490nm source. The exceptional brightness of the signal produced in dead cells makes SYTOX Green particularly useful for determining viability of not just mammalian cells, but both Gram-positive and Gram-negative bacteria.

Part of the Invitrogen Family of Cell Viability Stains and Assays
SYTOX stains for dead cells are available in a variety of colors, including red, blue, and orange. SYTOX stains have been incorporated into a number of assays for apoptosis, cell viability, and metabolism. 5mM solution in DMSO.

  • Impermeant to live cells
  • Excitation/Emission: 504/523nm
  • Use with 488 Argon-ion laser
  • Works with mammalian cells and both Gram-positive and Gram-negative bacteria

Cell Adhesion, Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, Microbial Tracking, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
504 nm
Tube(s)
250 μL
Fluorescence
Solution
Room Temperature
Nucleic Acid Stain
Store in freezer at -5°C to -30°C and protect from light.
Cell-Permeant
Flow Cytometer
250 μL
523 nm
SYTOX
Fluorescent Dye
Nucleic Acids, Nucleus
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2901546 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog NumberS7020
Lot Number2720299 Certificate TypeCertificate of Analysis Date19/09/2023 Catalog NumberS7020
Lot Number2521023 Certificate TypeCertificate of Analysis Date10/09/2022 Catalog NumberS7020
Lot Number2397782 Certificate TypeCertificate of Analysis Date02/12/2021 Catalog NumberS7020
Lot Number2306342 Certificate TypeCertificate of Analysis Date19/02/2021 Catalog NumberS7020
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SYTOX™ Green Nucleic Acid Stain - 5 mM Solution in DMSO > 250uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.