Learn More
Invitrogen™ TC-FlAsH™ II In-Cell Tetracysteine Tag Detection Kit (Green Fluorescence), for live-cell imaging
Contains an expression tag-based fluorescence labeling reagent for live-cell labeling
Brand: Invitrogen™ T34561
Code : 50
Additional Details : Weight : 0.02000kg
Includes: Kit contains FlAsH-EDT2 labeling reagent and BAL wash buffer.
Description
Mammalian cell lines expressing a protein fused to a tetracysteine tag (CCPGCC) can be labeled with the green-fluorescent FlAsH-EDT2 reagent contained in the kit. The tagged protein is fluorescent only when the labeling reagent is added. BAL wash buffer replaces the previously supplied Disperse Blue 3 and EDT-based wash buffer as an olfactorily more agreeable reagent that yields superior signal to noise.
Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Labeling Chemistry, Lumio Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays
Order Info
Shipping Condition: Wet Ice
Specifications
Tetracysteine Tag Detection Kit | |
Fluorescence Microscope | |
Store in freezer (-5 to -30°C) and protect from light. | |
FlAsH | |
Kit |
Fluorescent | |
Tetracysteine Tag Detection | |
Green | |
Wet Ice | |
TC-FlAsH II |
For Research Use Only. Not for use in diagnostic procedures.