Learn More
Invitrogen™ TMA-DPH (1-(4-Trimethylammoniumphenyl)-6-Phenyl-1,3,5-Hexatriene p-Toluenesulfonate)
For use with fluorescence microscopy
Brand: Invitrogen™ T204
456.95 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
TMA-DPH contains a cationic trimethylammonium substitute that acts as a surface anchor to improve the localization of the fluorescent probe of membrane interiors, DPH.
Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Endocytosis, Exocytosis & Phagocytosis, Membranes (General) & Lipids
Order Info
Shipping Condition: Room Temperature
Specifications
25 mg | |
Fluorescence Microscope | |
Fluorescence | |
Solid | |
Cell Surface Anchor |
Room Temperature | |
Plasma Membrane | |
C28H31NO3S | |
Violet |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.