Learn More
Invitrogen™ TNP-ATP (2'-(or-3')-O-(Trinitrophenyl) Adenosine 5'-Triphosphate, Trisodium Salt)
Structural probe
Brand: Invitrogen™ T7602
363.28 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The nucleotide analog TNP-ATP is modified on the ribose moiety and is essentially non-fluorescent in water. The TNP nucleotides undergo an equilibrium transition to a semiquinoid structure that has relatively long-wavelength spectral properties; this form is only fluorescent when bound to the nucleotide-binding site of some proteins.
Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Order Info
Shipping Condition: Wet ice
Specifications
TNP-ATP | |
Wet Ice | |
2 mL |
Store in freezer -5°C to -30°C and protect from light. | |
TNP |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.