Learn More
Invitrogen™ Molecular Probes™ TSA™ Kit 2, with HRP-Goat Anti-Mouse IgG and Alexa Fluor™ 488 Tyramide
Tyramide signal amplification is enzyme-mediated detection method that uses catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of target protein or nucleic acid sequence in situ
Brand: Invitrogen™ T20912
Code : 50
Additional Details : Weight : 0.11000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Includes: HRP-goat anti-mouse IgG and green-fluorescent Alexa Fluor 488 tyramide
Description
- Alexa Fluor 488 tyramide is spectrally similar to fluorescein
Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer (-5° to -30°C) and protect from light. |
For Research Use Only. Not for use in diagnostic procedures.